Learn More
Abeba™ Business Men 33240 Shoes
Product Code. 15629827
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15629827 43 1 pair
15659817 36 1 pair
15669817 37 1 pair
15679817 38 1 pair
15689817 39 1 pair
15699817 40 1 pair
15609827 41 1 pair
15619827 42 1 pair
15639827 44 1 pair
15649827 45 1 pair
15659827 46 1 pair
15669827 47 1 pair
15679827 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15629827 Supplier Abeba™ Supplier No. 33240/43
Forventet forsendelse: 10-12-2026
ESD dissipative with high wearing comfort. Smooth leather construction.
  • CE EN ISO 20345:2011 S2 SRA
  • Smooth leather
  • Black
  • Steel toe cap
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S2 SRA
Color Black
Liner Material Microfiber
Material Leather
Size 43
Sole Features Slip-resistant
Sole Material Rubber
Style 33240
Toe Material Steel