missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Busan 85, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Busan 85 |
| CAS | 128-03-0 |
| Molekylær formel | C3H6KNS2 |
| Mængde | 250 mg |
| Synonym | Dimethyldithiocarbamic Acid Potassium Salt (8CI), Dimethylcarbamodithioic Acid, Potassium Salt (9CI), Arylane, MBC 350, Potassium N,N-Dimethyldithiocarbamate, Potassium Dimethyldithiocarbamate |
| InChI formel | InChI=1S/C3H7NS2.K/c1-4(2)3(5)6;/h1-2H3,(H,5,6);/q;+1/p-1 |
| SMIL | [K+].CN(C)C(=S)[S-] |
| IUPAC navn | potassium;N,N-dimethylcarbamodithioate |
| Molekylvægt (g/mol) | 159.31 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?