Få mere at vide
Busan 85, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Busan 85 |
| CAS | 128-03-0 |
| Molekylær formel | C3H6KNS2 |
| Mængde | 100 mg |
| Synonym | Dimethyldithiocarbamic Acid Potassium Salt (8CI), Dimethylcarbamodithioic Acid, Potassium Salt (9CI), Arylane, MBC 350, Potassium N,N-Dimethyldithiocarbamate, Potassium Dimethyldithiocarbamate |
| InChI formel | InChI=1S/C3H7NS2.K/c1-4(2)3(5)6;/h1-2H3,(H,5,6);/q;+1/p-1 |
| SMIL | [K+].CN(C)C(=S)[S-] |
| IUPAC navn | potassium;N,N-dimethylcarbamodithioate |
| Molekylvægt (g/mol) | 159.31 |