Få mere at vide
Bupropion-d9 Hydrochloride, TRC
Artikelnummer. 30331240
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30331240 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30331240 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6896275MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (±)-Bupropion-D9 Hydrochloride
CAS 1189725-26-5
Renhedsgrad noter HPLC
Molekylær formel C13 2H9 H9 Cl N O . Cl H
Mængde 5 mg
Synonym 1-Propanone-3,3,3-d3, 1-(3-chlorophenyl)-2-[[1,1-di(methyl-d3)ethyl-2,2,2-d3]amino]-, hydrochloride (1:1), Bupropion-d9 Hydrochloride, (±)-Bupropion-D9 hydrochloride, Bupropion-D9 hydrochloride (N-tert-butyl D9)
InChI formel InChI=1S/C13H18ClNO.ClH/c1-9(15-13(2,3)4)12(16)10-6-5-7-11(14)8-10;/h5-9,15H,1-4H3;1H/i2D3,3D3,4D3;
SMIL Cl.[2H]C([2H])([2H])C(NC(C)C(=O)c1cccc(Cl)c1)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1-(3-chlorophenyl)-2-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]propan-1-one;hydrochloride
Molekylvægt (g/mol) 285.26
Formel vægt 284.1409
Procent renhed >95