Få mere at vide
Buprofezin, TRC
Artikelnummer. 30338331
Skift visning
Mængde:
250 mg
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30338331 25 g
30345583 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30338331 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68967025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Buprofezin
CAS 69327-76-0
Molekylær formel C16 H23 N3 O S
Mængde 25 g
Synonym 2-[(1,1-Dimethylethyl)imino]tetrahydro-3-(1-methylethyl)-5-phenyl-4H-1,3,5-thiadiazin-4-one, Applaud, Applaud 40SC, Buprofezin, Courier, NNI 750, Stater, Talus
InChI formel InChI=1S/C16H23N3OS/c1-12(2)19-14(17-16(3,4)5)21-11-18(15(19)20)13-9-7-6-8-10-13/h6-10,12H,11H2,1-5H3
SMIL CC(C)N1C(=NC(C)(C)C)SCN(C1=O)c2ccccc2
IUPAC navn 2-tert-butylimino-5-phenyl-3-propan-2-yl-1,3,5-thiadiazinan-4-one
Molekylvægt (g/mol) 305.44
Formel vægt 305.1562