Få mere at vide
Bupivacaine Hydrochloride, TRC
Artikelnummer. 30134576
Skift visning
Mængde:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134576 500 mg
30138608 5 g
30009655 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134576 Leverandør Toronto Research Chemicals Leverandørnr. TRCB780595500MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 18010-40-7
Molekylær formel C18 H28 N2 O . Cl H
Molekylvægt (g/mol) 324.89
Synonym 1-Butyl-N-(2,6-dimethylphenyl)-2-piperidinecarboxamide monohydrochloride, 1-Butyl-2',6'-pipecoloxylidide hydrochloride, 1-Butyl-2',6'-pipecoloxylidide monohydrochloride, (±)-Bupivacaine hydrochloride, AH 2250, Bicain, Bupivacaine hydrochloride, LAC 43, Marcaina, Marcaine hydrochloride, Sensorcaine, NovaBupi
IUPAC navn 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrochloride
SMIL Cl.CCCCN1CCCCC1C(=O)Nc2c(C)cccc2C
Tekniske data
Kemisk navn eller materiale Bupivacaine Hydrochloride
Mængde 500 mg
InChI formel InChI=1S/C18H28N2O.ClH/c1-4-5-12-20-13-7-6-11-16(20)18(21)19-17-14(2)9-8-10-15(17)3;/h8-10,16H,4-7,11-13H2,1-3H3,(H,19,21);1H
Formel vægt 324.1968