Få mere at vide
Bunolol Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Bunolol Hydrochloride |
| CAS | 31969-05-8 |
| Analyt- eller komponentnavne | Bunolol Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H25 N O3 . Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | 1(2H)-Naphthalenone, 5-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride (1:1), 1(2H)-Naphthalenone, 5-[3-[(1,1-dimethylethyl), 1(2H)-Naphthalenone, 5-[3-(tert-butylamino)-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride, (+/-)- (8CI), 1(2H)-Naphthalenone, 5-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride (9CI), 1(2H)-Naphthalenone, 5-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride, (+/-)-, (+/-)-Bunolol hydrochloride, Bunolol hydrochloride, W 6412A, dl-Bunolol hydrochloride |
| InChI formel | InChI=1S/C17H25NO3.ClH/c1-17(2,3)18-10-12(19)11-21-16-9-5-6-13-14(16)7-4-8-15(13)20;/h5-6,9,12,18-19H,4,7-8,10-11H2,1-3H3;1H |
| Løsningstype | Single Solution |
| SMIL | Cl.CC(C)(C)NCC(O)COc1cccc2C(=O)CCCc12 |
| Vis mere |