Learn More
Brady™ Bulk Rapido Wire Marking Tags for M710, M610, M611
Bulk Rapido™ Wire Marking tags for use with Brady™ Printers M611, M610 and M710.
Brand: Brady™ 312138
Additional Details : Weight : 0.58400kg
Description
- Rapido™ Wiremarking Sleeves are fast and easy to apply
- Simply squeeze the edges of the material together, thread your wire through the holes and pull away the wire to release the marker
- Provides high tear, smudge and abrasion resistance
Specifications
250 Labels | |
RoHS Directive 2011/65/EU | |
Polyethylene | |
Data and Telecommunications Labeling, Wire and Cable Labeling | |
0.205 in. | |
0.25 mm | |
-40°F to +122°F | |
23 mm | |
3.5 mm |
Tag | |
Yellow | |
Matte | |
5.2 mm | |
Rectangle | |
-40°C to +50°C | |
0.906 in. | |
0.138 in. | |
M610, M710, M611, BMP61, BMP71 |