Få mere at vide
Bufuralol-d9 Hydrochloride, TRC
Artikelnummer. 30347741
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30347741 10 mg
30330649 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30347741 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68954210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bufuralol-d9 Hydrochloride
CAS 1173023-51-2
Molekylær formel C16H15D9ClNO2
Mængde 10 mg
Synonym α-[[(1,1-Dimethylethyl)-amino]methyl]-7-ethyl-2-benzofuranmethanol-d9, Ro-3-4787-d9, Angium-d9
InChI formel InChI=1S/C16H23NO2.ClH/c1-5-11-7-6-8-12-9-14(19-15(11)12)13(18)10-17-16(2,3)4;/h6-9,13,17-18H,5,10H2,1-4H3;1H/i2D3,3D3,4D3;
SMIL OC(CNC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C(O1)=CC2=C1C(CC)=CC=C2.Cl
IUPAC navn 1-(7-ethyl-1-benzofuran-2-yl)-2-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]ethanol;hydrochloride
Molekylvægt (g/mol) 306.88