Få mere at vide
Buformin Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Buformin Hydrochloride |
| CAS | 1190-53-0 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C6 H15 N5 . Cl H |
| Mængde | 25 mg |
| Synonym | Biguanide, 1-butyl-, monohydrochloride, Imidodicarbonimidic diamide, N-butyl-, monohydrochloride, 1-Butylbiguanide hydrochloride, 1-Butylbiguanide, monohydrochloride, 1-Butyldiguanide hydrochloride, Andere, Biforon, Bigunal, Bufonamin, Buformin hydrochloride, Bulbonin, Diabrin, Dibetos, Glibutide, Gliporal, Glybutide, Insulamin, Krebon, N-Butylbiguanide hydrochloride, N1-Butyl-biguanide hydrochloride, N1-n-Butylbiguanide hydrochloride, Panformin, Sindiatil, Tidemol, Ziavetine |
| InChI formel | InChI=1S/C6H15N5.ClH/c1-2-3-4-10-6(9)11-5(7)8;/h2-4H2,1H3,(H6,7,8,9,10,11);1H |
| SMIL | Cl.CCCCNC(=N)NC(=N)N |
| IUPAC navn | 1-butyl-3-carbamimidoylguanidine;hydrochloride |
| Molekylvægt (g/mol) | 193.68 |
| Vis mere |