missing translation for 'onlineSavingsMsg'
Få mere at vide

Buformin-d9 Hydrochloride, TRC

Artikelnummer. 30342172
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30342172 1 mg
30337583 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30342172 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6895221MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Buformin-d9 Hydrochloride
CAS 1215700-80-3
Molekylær formel C6H7D9ClN5
Mængde 1 mg
Synonym N-Butyl-d9-idodicarbonimidic Diamide Hydrochloride, 1-Butyl-d9-biguanide Hydrochloride, 1-Butylbiguanide-d9 Hydrochloride, Buformine-d9 Hydrochloride, Butformin-d9 Hydrochloride, Butylbiguanide-d9 Hydrochloride, Butyldiguanide-d9 Hydrochloride, DBV-d9 Hydrochloride, Dibetos B-d9 Hydrochloride, Glybigid-d9 Hydrochloride, H 224-d9 Hydrochloride, N1-Butylbiguanide-d9 Hydrochloride, W 37-d9 Hydrochloride
InChI formel InChI=1S/C6H15N5.ClH/c1-2-3-4-10-6(9)11-5(7)8;/h2-4H2,1H3,(H6,7,8,9,10,11);1H/i1D3,2D2,3D2,4D2;
SMIL NC(NC(NC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=N)=N.Cl
IUPAC navn 3-Carbamimidoyl-1-(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)guanidine hydrogen chloride
Molekylvægt (g/mol) 202.73
Formel vægt 202.17
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.