Få mere at vide
Buflomedil Hydrochloride, LoGiCal
Artikelnummer. 30459421
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30459421 10 mg
1 options
Artikelnummer. 30459421 Leverandør LoGiCal Leverandørnr. LGCFOR0066.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Buflomedil hydrochloride
CAS 35543-24-9
Analyt- eller komponentnavne Buflomedil hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C17 H25 N O4 . Cl H
Certificeringer/compliance ISO 9001
Synonym Buflomedil Hydrochloride, 4-(1-Pyrrolidinyl)-1-(2,4,6-trimethoxyphenyl)-1-butanone hydrochloride, Bufedil, Buflan, Buflocit, Buflomedil hydrochloride, Buflonat, Fonzylane, Irrodan, Irrodan R, LL 1656, Lofton, Loftyl, Provas
InChI formel InChI=1S/C17H25NO4.ClH/c1-20-13-11-15(21-2)17(16(12-13)22-3)14(19)7-6-10-18-8-4-5-9-18;/h11-12H,4-10H2,1-3H3;1H
Løsningstype Neat
SMIL Cl.COc1cc(OC)c(C(=O)CCCN2CCCC2)c(OC)c1
IUPAC navn 4-pyrrolidin-1-yl-1-(2,4,6-trimethoxyphenyl)butan-1-one;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 343.85
Mængde 10 mg
Formel vægt 343.155 g/mol