missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 4, Scharlab

Potassium hydrogen phthalate buffer for use in calibrating pH meters in analytical chemistry.

Brand:  Scharlab SO10041000

Quantity 1 L

 View more versions of this product

Product Code. 11359893

  • 213.00 DKK / 1 L

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Description

Description

Standard buffer solutions are prepared using gravimetric and volumetric procedures. The batch value is determined by measurement with a combination glass electrode against five-point calibration according to DIN 19268. This pH buffer solution is traceable to Standard Reference Material from NIST.

Specifications

Chemical Identifiers

Potassium hydrogen phthalate buffer

Specifications

1.01g/cm3
Solution
Per liter: 10.21g Potassium hydrogen phthalate. Contains preservative.
Potassium hydrogen phthalate buffer
4.00
1 L
Traceable to NIST
Miscible in water (20°C)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Solution pH 4, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.