missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution for HPCE, 20 mM sodium phosphate, pH 8.5±0.1 (25 °C), Honeywell Fluka™

Brand:  Honeywell Fluka 82601-50ML

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 15637220

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +45 70 27 99 20 for assistance


Specifications

Specifications

1: 0 to 0.02; 2: 0 to 0.04; 3: 0 to 0.12; 4: 0 to 1.2
UV absorption/200 to 800nm in accordance
Liquid
20mM sodium phosphate
Plastic bottle
50mL
Buffer solution HPCE pH 8.5
Colorless
2.5 to 3.5mS/cm at 20°C
NONH for all modes of transport
8.5±0.1 (25°C)
In accordance for HPCE
Videos