missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution for HPCE, 100 mM sodium phosphate, pH 2.5 (25 °C), Honeywell Fluka™

Brand:  Honeywell Fluka 82578-500ML

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15637180

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +45 70 27 99 20 for assistance


Specifications

Specifications

Buffer solution HPCE pH 2.5
Colorless
6mS/cm
NONH for all modes of transport
2.5 (25°C)
In accordance for HPCE
UV absorption/200 to 800nm in accordance
Liquid
100mM sodium phosphate
Plastic bottle
500mL
Videos