missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution, Cs2CO3 for AAS 0.02mg/L HNO3 matrix, Fisher chemical™
Shop All Fisher Chemical Products

Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Specifications
Specifications
Concentration | 0.02 mg/L |
Solvent or Matrix | HNO3 matrix |
Quantity | 100 mL |
Chemical Name or Material | Buffer solution, Cs2CO3 for AAS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer solution, Cs2CO3 for AAS 0.02mg/L HNO3 matrix, Fisher chemical™ >
Spot an opportunity for improvement?Share a Content Correction