missing translation for 'onlineSavingsMsg'
Learn More
Buffer Concentrated Solution pH8.00 (Borate), NIST Standard for pH measurement, Fisher Chemical™ Product Code.: 10764074

Buffer Concentrated Solution pH8.00 (Borate), NIST Standard for pH measurement, Fisher Chemical™

Product Code. p-7073119 Shop All Fisher Chemical Products
1 Ampule, 1 ampoule
Click to view available options
Quantity:
1 Ampule
6 Ea.
Unit Size:
1 ampoule
Pack of 6
This item is not returnable. View return policy

Product Code. 10764074

Brand: Fisher Chemical J/2860C/05

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

MDL Number 163147

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 Ampule
Packaging Ampoule

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot Number2535301 Certificate TypeCertificate of Analysis Date24/05/2025 Product CodeJ/2860C/90,J/2860/17,J/2860/08,J/2860/24,J/2860C/05,J/2860/15
Lot Number2529548 Certificate TypeCertificate of Analysis Date24/05/2025 Product CodeJ/2860C/90,J/2860/17,J/2860/08,J/2860/24,J/2860C/05,J/2860/15
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Concentrated Solution pH8.00 (Borate), NIST Standard for pH measurement, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.