missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Colour Coded Concentrated Solution pH7.00 (Phosphate) Yellow, NIST Standard for pH measurement, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 Ampule
6 Ea.
Unit Size:
1 ampoule
Pack of 6
Specifications
Specifications
Color | Yellow |
Physical Form | Liquid |
Quantity | 1 Ampule |
Packaging | Ampoule |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
2528161 | Certificate of Analysis | 24/05/2025 | J/2855/15,J/2855C/05,J/2855C/90,J/2855/17,J/2855/08,J/2855/21,J/2855/24 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer Colour Coded Concentrated Solution pH7.00 (Phosphate) Yellow, NIST Standard for pH measurement, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction