Få mere at vide
Bufexamac, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Bufexamac |
| CAS | 2438-72-4 |
| Molekylær formel | C12 H17 N O3 |
| Mængde | 5 g |
| Synonym | Acetohydroxamic acid, 2-(p-butoxyphenyl)- (7CI,8CI), 4-Butoxy-N-hydroxybenzeneacetamide, 2-(p-Butoxyphenyl)acetohydroxamic acid, 4-Butoxyphenylacethydroxamic acid, Anderm, Bufexamac, Bufexamic acid, CP 1044J3, Droxarol, Droxaryl, Feximac, Flogocid, Flogocid N plastigel, Malipuran, Mofenar, Norfemac, Parfenac, Parfenal, Trolab, p-Butoxyphenylacethydroxamic acid, p-Butoxyphenylacetohydroxamic acid |
| InChI formel | InChI=1S/C12H17NO3/c1-2-3-8-16-11-6-4-10(5-7-11)9-12(14)13-15/h4-7,15H,2-3,8-9H2,1H3,(H,13,14) |
| SMIL | CCCCOc1ccc(CC(=O)NO)cc1 |
| IUPAC navn | 2-(4-butoxyphenyl)-N-hydroxyacetamide |
| Molekylvægt (g/mol) | 223.27 |
| Formel vægt | 223.1208 |