Få mere at vide
Bufexamac, Mikromol™
Beskrivelse
Bufexamac
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Bufexamac |
| CAS | 2438-72-4 |
| Analyt- eller komponentnavne | Bufexamac |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C12 H17 N O3 |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Bufexamac, 4-Butoxy-N-hydroxy-benzeneacetamide, 2-(p-Butoxyphenyl)-acetohydroxamic acid, 2-(p-Butoxyphenyl)acetohydroxamic acid, 4-Butoxyphenylacethydroxamic acid, Anderm, Bufexamic acid, CP 1044J3, Droxarol, Droxaryl, Feximac, Flogocid, Flogocid N plasti, Flogocid N plastigel, Malipuran, Mofenar, Norfemac, Parfenac, Parfenal, p-Butoxyphenylacethydroxamic acid, p-Butoxyphenylacetohydroxamic acid |
| InChI formel | InChI=1S/C12H17NO3/c1-2-3-8-16-11-6-4-10(5-7-11)9-12(14)13-15/h4-7,15H,2-3,8-9H2,1H3,(H,13,14) |
| SMIL | CCCCOc1ccc(CC(=O)NO)cc1 |
| IUPAC navn | 2-(4-butoxyphenyl)-N-hydroxyacetamide |
| Vis mere |