Få mere at vide
Bufexamac, LoGiCal
Artikelnummer. 30462581
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30462581 10 mg
1 options
Artikelnummer. 30462581 Leverandør LoGiCal Leverandørnr. LGCFOR0179.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bufexamac
CAS 2438-72-4
Analyt- eller komponentnavne Bufexamac
Forsendelsestilstand Room Temperature
Molekylær formel C12 H17 N O3
Certificeringer/compliance ISO 9001
Synonym Bufexamac, 4-Butoxy-N-hydroxy-benzeneacetamide, 2-(p-Butoxyphenyl)-acetohydroxamic acid, 2-(p-Butoxyphenyl)acetohydroxamic acid, 4-Butoxyphenylacethydroxamic acid, Anderm, Bufexamic acid, CP 1044J3, Droxarol, Droxaryl, Feximac, Flogocid, Flogocid N plasti, Flogocid N plastigel, Malipuran, Mofenar, Norfemac, Parfenac, Parfenal, p-Butoxyphenylacethydroxamic acid, p-Butoxyphenylacetohydroxamic acid
InChI formel InChI=1S/C12H17NO3/c1-2-3-8-16-11-6-4-10(5-7-11)9-12(14)13-15/h4-7,15H,2-3,8-9H2,1H3,(H,13,14)
Løsningstype Neat
SMIL CCCCOc1ccc(CC(=O)NO)cc1
IUPAC navn 2-(4-butoxyphenyl)-N-hydroxyacetamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 223.27
Mængde 10 mg
Formel vægt 223.1208 g/mol