Få mere at vide
Budesonide-d8, TRC
Artikelnummer. 30339135
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30339135 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30339135 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68949210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Budesonide-d8
CAS 1105542-94-6
Renhedsgrad noter HPLC
Molekylær formel C25H26D8O6
Mængde 10 mg
Synonym (11β,16α)-16,17-[Butylidenebis(oxy)]-11,21-dihydroxypregna-1,4-diene-3,20-dione, Bidien, Budenofalk, Budesomide, Budeson, Cortivent, Entocort, Micronyl, Nebuampul, Preferid, Pulmicort, Respules, Rhinocort, Rhinocort Aqua, S 1320, Spirocort, Turbuhaler
InChI formel InChI=1S/C25H34O6/c1-4-5-21-30-20-11-17-16-7-6-14-10-15(27)8-9-23(14,2)22(16)18(28)12-24(17,3)25(20,31-21)19(29)13-26/h8-10,16-18,20-22,26,28H,4-7,11-13H2,1-3H3/t16-,17-,18-,20+,21?,22+,23-,24-,25+/m0/s1/i1D3,4D2,5D2,21D
SMIL O=C1C=C[C@@]2(C)C(CC[C@]3([H])[C@]2([H])[C@@H](O)C[C@@]4(C)[C@@]3([H])C[C@@H](OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])([2H])O5)[C@]45C(CO)=O)=C1
IUPAC navn (1S,2S,4R,8S,9S,11S,12S,13R)-6-deuterio-6-(1,1,2,2,3,3,3-heptadeuteriopropyl)-11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-16-one
Molekylvægt (g/mol) 438.58
Procent renhed >95