Få mere at vide
Budesonide-d6, TRC
Artikelnummer. 30347807
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30347807 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30347807 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68949310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Budesonide-d6
CAS 1134189-63-1
Molekylær formel C25H28D6O6
Mængde 10 mg
Synonym (11β,16α)-16,17-[Butylidenebis(oxy)]-11,21-dihydroxypregna-1,4-diene-3,20-dione-d6, Bidien-d6, Budenofalk-d6, Budesomide-d6, Budeson-d6, Cortivent-d6, Entocort-d6, Micronyl-d6, Nebuampul-d6, Preferid-d6, Pulmicort-d6, Respules-d6, Rhinocort-d6, Rhinocort Aqua-d6-d6, S 1320-d6, Spirocort-d6, Turbuhaler-d6
InChI formel InChI=1S/C25H34O6/c1-4-5-21-30-20-11-17-16-7-6-14-10-15(27)8-9-23(14,2)22(16)18(28)12-24(17,3)25(20,31-21)19(29)13-26/h8-10,16-18,20-22,26,28H,4-7,11-13H2,1-3H3
SMIL O=C1C([2H])=C[C@@]2(C)C(C([2H])([2H])C[C@]3([H])[C@]2([H])[C@H](O)C[C@@]4(C)[C@]3([H])C[C@@H](OC(CCC)O5)[C@]45C(C([2H])([2H])O)=O)=C1[2H]
IUPAC navn 11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-16-one
Molekylvægt (g/mol) 436.57
Formel vægt 436.2732