Få mere at vide
Buclosamide, LoGiCal
Artikelnummer. 30458150
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30458150 5 mg
1 options
Artikelnummer. 30458150 Leverandør LoGiCal Leverandørnr. LGCFOR1842.00
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Buclosamide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Buclosamide
CAS 575-74-6
Analyt- eller komponentnavne Buclosamide
Forsendelsestilstand Room Temperature
Molekylær formel C11 H14 Cl N O2
Certificeringer/compliance ISO 9001
Synonym Benzamide, N-butyl-4-chloro-2-hydroxy-, Salicylamide, N-butyl-4-chloro- (6 CI,7 CI,8 CI), 4-Chloro-2-hydroxybenzoic acid-n-butylamide, Buclosamide, Demycin, Fungit, N-Butyl-4-chloro-2-hydroxybenzamide, N-Butyl-4-chlorosalicylamide
InChI formel InChI=1 S/C11H14ClNO2/c1-2-3-6-13-11(15)9-5-4-8(12)7-10(9)14/h4-5,7,14 H,2-3,6H2,1H3,(H,13,15)
SMIL CCCCNC(=O)c1ccc(Cl)cc1O
IUPAC navn N-butyl-4-chloro-2-hydroxybenzamide
Industry Type Forensics
Molekylvægt (g/mol) 227.69
Mængde 5 mg
Formel vægt 227.0713 g/mol