Learn More
Thermo Scientific™ Buckets for Thermo Scientific™ HAEMAFlex™ Swinging Bucket Rotors
Product Code. 15880582 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
HAEMAFlex 12/16 Swing Bucket Rotor
HAEMAFlex 6/8 Swing Bucket Rotor
Description:
Double Bucket
Oval Single Bucket
Round Bucket
Unit Size:
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Description Unit Size
15880582 HAEMAFlex 6/8 Swing Bucket Rotor Round Bucket Pack of 2
15600937 HAEMAFlex 12/16 Swing Bucket Rotor Double Bucket Pack of 2
15729389 HAEMAFlex 6/8 Swing Bucket Rotor Oval Single Bucket Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15880582 Supplier Thermo Scientific™ Supplier No. 75003836
  • ONLINE EXCLUSIVE PRICE
    12160.00 DKK
    11489.88 DKK / Pakke med 2
    Spare 670.12 DKK
    5% Slukket
Forventet forsendelse: 05-03-2027
Expand individual bucket capacity or maximize total blood bags per spin with choice of buckets for Thermo Scientific™ HAEMAFlex™ blood banking rotors.
Thermo Scientific blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Address your lab’s regular- to medium-throughput blood processing needs with the HAEMAFlex 6 swinging bucket rotor with 6 x 550mL blood bag capacity or, utilizing the same bucket system, upgrade to 8 x 550mL bags with the HAEMAFlex 8 swinging bucket rotor.
For high-throughput blood processing needs, the HAEMAFlex 12 swinging bucket rotor provides 12 x 500mL capacity or upgrade your centrifuge capacity to 16 x 500mL blood bags with the HAEMAFlex 16 rotor and the same bucket selections.
Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) HAEMAFlex 6/8 Swing Bucket Rotor
Capacity 16 x 500 mL
Product Type Single Blood Bag Bucket
Description Round Bucket