Learn More
Grathwol Bucket with Lid
Used for storing purposes.
POA - 18.62 DKK
Product Code | Brand | Height (Metric) | Diameter (Metric) Outer | Capacity (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Diameter (Metric) Outer | Capacity (Metric) | Price | Quantity & Availability | |||
12317647
|
Grathwol
321860040 |
197 mm | 200 mm | 5.8 L |
18.62 DKK
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
12307647
|
Grathwol
321706040 |
396 mm | 259 mm | 21.7 L |
POA
Each |
N/A | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
Description
- Used for storing purposes
Specifications
Bucket with Lid |