Learn More
BUCHI Condenser Module for V-710 Pump
Secondary condenser V Vertical condenser, includes insulator. Condenses remaining vapors after the vacuum pump outlet. Used along with a recirculating chiller or tap water.
Brand:  BUCHI 047180
Additional Details : Weight : 0.98000kg
Product Code. 10190573
  • 11680.00 DKK / Each
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
Description
Description
  • Ensure all solvent vapors are contained before they can reach the environment
  • Especially useful for low-boiling solvents
  • Add a level of security for enclosed spaces
  • Content: 500 mL receiving flask, insulator for condenser, ball joint clamp, holder
Specifications
Specifications
V-710 vacuum pump
Mounting brackets, tubing connections and receiving flask