Learn More
BUCHI Condenser Module for V-710 Pump
Secondary condenser V Vertical condenser, includes insulator. Condenses remaining vapors after the vacuum pump outlet. Used along with a recirculating chiller or tap water.
Brand: BUCHI 047180
Additional Details : Weight : 0.98000kg
Description
- Ensure all solvent vapors are contained before they can reach the environment
- Especially useful for low-boiling solvents
- Add a level of security for enclosed spaces
- Content: 500 mL receiving flask, insulator for condenser, ball joint clamp, holder
Specifications
V-710 vacuum pump | |
Mounting brackets, tubing connections and receiving flask |