missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Novus Biologicals™ BTLA/CD272 Antibody (icatolimab) - Humanized, IgG4SP, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28499-1mg
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
BTLA/CD272 Monoclonal antibody specifically detects BTLA/CD272 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Tekniske data
| BTLA/CD272 | |
| Monoclonal | |
| Unconjugated | |
| Q7Z6A9 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| icatolimab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 | |
| BTLA/ CD272 | |
| 1 mg | |
| Cell Cycle and Replication, Phospho Specific | |
| 151888 | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion