Få mere at vide
BTF Antibody [HRP], Novus Biologicals Biologicals™
Artikelnummer. 30498874 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mL
Artikelnummer. Mængde
30498874 0.1 mL
1 options
Artikelnummer. 30498874 Leverandør Novus Biologicals Leverandørnr. NBP327902H
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Rabbit Polyclonal Antibody
BTF Polyclonal antibody specifically detects BTF in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Tekniske data
Antigen BTF
Applikationer ELISA, Western Blot, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugeret HRP
Formulering PBS
Gene Alias bcl-2-associated transcription factor 1, BCL2-associated transcription factor 1, Btf, BTFKIAA0164bK211L9.1
Værtsarter Rabbit
Immunogen BTF antibody was raised against a 19 amino acid peptide near the carboxy terminus of human BTF. The immunogen is located within amino acids 820 - 870 of BTF.
Oprensningsmetode Peptide affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Apoptosis, DNA Repair
Primær eller sekundær Primary
Gen-id (Entrez) 9774
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at 4°C in the dark.
Produkttype Antibody
Form Purified
Isotype IgG