Få mere at vide
Tocris Bioscience™ BTD
Beskrivelse
BTD is a selective TRPC5 activator (EC50 = 1.4 μM in a calcium influx assay, 1.3 μM in whole cell patch clamp assay). Displays >14.5-fold selectivity for TRPC5 over other TRP channels (EC50 = 20.6 μM for TRPM8). Also activates TRPC1:5 and TRPC4:5 heteromers.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 6-Methyl-N-[3-(tricyclo[3.3.1.13,7]dec-1-yloxy)propyl]-4H-1,2,4-benzothiadiazine-3-propanamide-1,1-dioxide |
| CAS | 896684-04-1 |
| Mængde | 50 mg |
| Molekylær formel | C24H33N3O4S |
| Renhed | 98% |