missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brucine sulfate hydrate, 98% (dry wt.), water <13%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Brucine sulfate hydrate is used as a tool for stereospecific chemical syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Brucine sulfate hydrate is used as a tool for stereospecific chemical syntheses.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Brucine sulfate hydrate is used as a tool for stereospecific chemical syntheses.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
Melting Point | 191°C to 196°C (decomposition) |
Quantity | 25 g |
UN Number | UN2811 |
Sensitivity | Hygroscopic |
Merck Index | 14,1455 |
Solubility Information | Soluble in water. |
Formula Weight | 443.52 (Anhydrous) |
Percent Purity | 98% |
Assay | (dry wt.), water <13% |
Chemical Name or Material | Brucine sulfate hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'