Få mere at vide
Tocris Bioscience™ Bropirimine
GREENER_CHOICE
Artikelnummer. 18033812
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18033812 10 mg
18084852 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18033812 Leverandør Tocris Bioscience™ Leverandørnr. 4316/10
Immunomodulatory and antitumor compound
Immunomodulatory and antitumor compound. Induces interferon (IFN)-α/β production; augments natural killer (NK) cell activity. Orally active.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 56741-95-8
Molekylær formel C10H8BrN3O
Tekniske data
Kemisk navn eller materiale Bropirimine
Mængde 10 mg
Formel vægt 266.09
Renhed >99%
Molekylvægt (g/mol) 266.098