Learn More
Bromotrimethylsilane, Sigma-Aldrich™
Product Code. 15814311
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Product Code. Quantity Unit Size
15814311 10 mL 10mL
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15814311 Supplier Sigma Aldrich Supplier No. 8143240010
 Restricted Item
Forventet forsendelse: 26-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 2857-97-8
Melting Point -43°C
Density 1.17 g/cm3 at 20°C
Boiling Point 79°C/1013 hPa
Molecular Formula (CH3)3SiBr
MDL Number MFCD00000048
Quantity 10 mL
Synonym Bromotrimethylsilane, Trimethylbromosilane, Trimethylsilyl bromide, TMB, TMBS, Bromotrimethylsilane
Molecular Weight (g/mol) 153.09
Grade Research