missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromophenol Blue sodium salt, 0.04% w/v aq. soln., Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
6 x 100 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Concentration | 0.04% w/v aq. soln. |
Merck Index | 141444 |
Formula Weight | 691.96 |
Color | Red |
Quantity | 100 mL |
Physical Form | Liquid |
Chemical Name or Material | Bromophenol Blue sodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'