missing translation for 'onlineSavingsMsg'
Learn More

Bromopentafluoroacetone, 95%, Thermo Scientific Chemicals

Catalog Number 11363247
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
815-23-6
C3BrF5O
226.928
MFCD00153080
IYDJQZOFWOSTFO-UHFFFAOYSA-N
bromopentafluoroacetone, bromopentafluoro-2-propanone, bromopentafluoropropan-2-one, 1-bromo-1,1,3,3,3-pentafluoroacetone, 3-bromo-1,1,1,3,3-pentafluoroacetone, 1-bromo-1,1,3,3,3-pentafluoro-2-propanone, 2-propanone,1-bromo-1,1,3,3,3-pentafluoro, 1-bromanyl-1,1,3,3,3-pentakis fluoranyl propan-2-one
2736352
1-bromo-1,1,3,3,3-pentafluoropropan-2-one
C(=O)(C(F)(F)F)C(F)(F)Br
This item is not returnable. View return policy
815-23-6
C3BrF5O
226.928
MFCD00153080
IYDJQZOFWOSTFO-UHFFFAOYSA-N
bromopentafluoroacetone, bromopentafluoro-2-propanone, bromopentafluoropropan-2-one, 1-bromo-1,1,3,3,3-pentafluoroacetone, 3-bromo-1,1,1,3,3-pentafluoroacetone, 1-bromo-1,1,3,3,3-pentafluoro-2-propanone, 2-propanone,1-bromo-1,1,3,3,3-pentafluoro, 1-bromanyl-1,1,3,3,3-pentakis fluoranyl propan-2-one
2736352
1-bromo-1,1,3,3,3-pentafluoropropan-2-one
C(=O)(C(F)(F)F)C(F)(F)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 31°C to 33°C
Quantity 1 g
UN Number UN3265
Beilstein 1773590
Sensitivity Light sensitive
Formula Weight 226.93
Percent Purity 95%
Chemical Name or Material Bromopentafluoroacetone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'