Få mere at vide
Bromomethyl Methanethiosulfonate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Bromomethyl Methanethiosulfonate |
| CAS | 887353-78-8 |
| Molekylær formel | C2 H5 Br O2 S2 |
| Mængde | 250 mg |
| Synonym | Methanesulfonothioic acid, S-(bromomethyl) ester, Bromo-methylsulfonylsulfanylmethane, Bromomethyl methanethiosulfonate, Methanesulfonothioic acid S-(bromomethyl) ester |
| InChI formel | InChI=1S/C2H5BrO2S2/c1-7(4,5)6-2-3/h2H2,1H3 |
| SMIL | CS(=O)(=O)SCBr |
| IUPAC navn | bromo(methylsulfonylsulfanyl)methane |
| Molekylvægt (g/mol) | 205.09 |
| Formel vægt | 203.8914 |