Få mere at vide
Bromoethane-d3, TRC
Artikelnummer. 30341944
Skift visning
Mængde:
5 g
Artikelnummer. Mængde
30341944 5 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30341944 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6841985G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bromoethane-d3
CAS 7439-86-3
Molekylær formel C2 D3 H2 Br
Mængde 5 g
Synonym Ethyl Bromide-d3, 2-Bromoethane-2,2,2-d3, Bromoethane-d3, Ethyl-d3 Bromide, Trideuterioethyl Bromide-d3, 2,2,2-Trideuteroethyl Bromide, 2-Bromo-1,1,1-trideuteroethane
InChI formel InChI=1S/C2H5Br/c1-2-3/h2H2,1H3/I1d3
SMIL [2H]C([2H])([2H])CBr
IUPAC navn 2-bromo-1,1,1-trideuterioethane
Molekylvægt (g/mol) 111.98
Formel vægt 110.9763