Få mere at vide
Bromoethane-d3, TRC
Artikelnummer. 30348218
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30348218 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30348218 Leverandør Toronto Research Chemicals Leverandørnr. TRCB684198250MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bromoethane-d3
CAS 7439-86-3
Molekylær formel C2 D3 H2 Br
Mængde 250 mg
Synonym Ethyl Bromide-d3, 2-Bromoethane-2,2,2-d3, Bromoethane-d3, Ethyl-d3 Bromide, Trideuterioethyl Bromide-d3, 2,2,2-Trideuteroethyl Bromide, 2-Bromo-1,1,1-trideuteroethane
InChI formel InChI=1S/C2H5Br/c1-2-3/h2H2,1H3/I1d3
SMIL [2H]C([2H])([2H])CBr
IUPAC navn 2-bromo-1,1,1-trideuterioethane
Molekylvægt (g/mol) 111.98
Formel vægt 110.9763