Få mere at vide
Bromoethane-2-d1, CDN
Artikelnummer. 30321364
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321364 1 g -
30317071 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321364 Leverandør CDN Leverandørnr. CDND03441G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromoethane-2-D
CAS 23705-67-1
Molekylær formel C2 D H4 Br
Mængde 1 g
Synonym Ethane-d, 2-bromo- (6 CI, 8 CI, 9 CI), 1-Bromoethane-2-d1, 2-Deuteroethyl bromide, 2-Bromoethane-1-d, 1-Bromoethane-2-D
InChI formel InChI=1 S/C2H5Br/c1-2-3/h2H2,1H3/i1D
SMIL [2 H]CCBr
IUPAC navn 1-bromo-2-deuterioethane
Molekylvægt (g/mol) 109.9713
Formel vægt 108.9637 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity