Få mere at vide
Bromoethane-2,2,2-d3, CDN
Artikelnummer. 30319232
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319232 1 g
30311081 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319232 Leverandør CDN Leverandørnr. CDND03071G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bromoethane-d3
CAS 7439-86-3
Molekylær formel C2 D3 H2 Br
Mængde 1 g
Synonym Ethyl Bromide-d3, 2-Bromoethane-2,2,2-d3;Bromoethane-d3, Ethyl-d3 Bromide, Trideuterioethyl Bromide-d3, 2,2,2-Trideuteroethyl Bromide, 2-Bromo-1,1,1-trideuteroethane
InChI formel InChI=1 S/C2H5Br/c1-2-3/h2H2,1H3/I1d3
SMIL [2 H]C([2 H])([2 H])CBr
IUPAC navn 2-bromo-1,1,1-trideuterioethane
Molekylvægt (g/mol) 111.98
Formel vægt 110.9763 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity