Få mere at vide
Bromodichloroacetonitrile, TRC
Artikelnummer. 30133387
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133387 500 mg
17903275 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133387 Leverandør Toronto Research Chemicals Leverandørnr. TRCB683400500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 60523-73-1
Molekylær formel C2 Br Cl2 N
Molekylvægt (g/mol) 188.84
Synonym 2-Bromo-2,2-dichloroacetonitrile;
IUPAC navn 2-bromo-2,2-dichloroacetonitrile
SMIL ClC(Cl)(Br)C#N
Tekniske data
Kemisk navn eller materiale Bromodichloroacetonitrile
Mængde 500 mg
InChI formel InChI=1S/C2BrCl2N/c3-2(4,5)1-6
Formel vægt 186.8591