Få mere at vide
Bromocyclopropane-d5, CDN
Artikelnummer. 30319121
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319121 0.25 g -
30315634 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319121 Leverandør CDN Leverandørnr. CDND77420.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bromocyclopropane-d5
CAS 1643577-64-3
Molekylær formel C3 D5 Br
Mængde 0.25 g
Synonym Cyclopropyl bromide-d5, NSC 89692-d5
InChI formel InChI=1 S/C3H5Br/c4-3-1-2-3/h3H,1-2H2/i1D2,2D2,3 D
SMIL [2 H]C1([2 H])C([2 H])([2 H])C1([2 H])Br
IUPAC navn 1-bromo-1,2,2,3,3-pentadeuteriocyclopropane
Molekylvægt (g/mol) 126.01
Formel vægt 124.9888 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity