Få mere at vide
Bromocyclopropane-d4, TRC
Artikelnummer. 30339309
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30339309 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30339309 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68276225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bromocyclopropane-d4
CAS 1398065-55-8
Molekylær formel C3 D4 H Br
Mængde 25 mg
Synonym Cyclopropyl bromide-d4, NSC 89692-d4
InChI formel InChI=1S/C3H5Br/c4-3-1-2-3/h3H,1-2H2/i1D2,2D2
SMIL [2H]C1([2H])C(Br)C1([2H])[2H]
IUPAC navn 3-bromo-1,1,2,2-tetradeuteriocyclopropane
Molekylvægt (g/mol) 125
Formel vægt 123.9826