missing translation for 'onlineSavingsMsg'
Få mere at vide

Bromocyclopropane-d4, TRC

Artikelnummer. 30344013
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30344013 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30344013 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68276250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Kemisk navn eller materiale Bromocyclopropane-d4
CAS 1398065-55-8
Molekylær formel C3 D4 H Br
Mængde 50 mg
Synonym Cyclopropyl bromide-d4, NSC 89692-d4
InChI formel InChI=1S/C3H5Br/c4-3-1-2-3/h3H,1-2H2/i1D2,2D2
SMIL [2H]C1([2H])C(Br)C1([2H])[2H]
IUPAC navn 3-bromo-1,1,2,2-tetradeuteriocyclopropane
Molekylvægt (g/mol) 125
Formel vægt 123.9826
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.