Få mere at vide
Bromocyclopentane-d9, TRC
Artikelnummer. 30332735
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30332735 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30332735 Leverandør Toronto Research Chemicals Leverandørnr. TRCB682827250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bromocyclopentane-d9
CAS 35468-44-1
Molekylær formel C5 D9 Br
Mængde 250 mg
Synonym 5-Bromo-cyclopentane-1,1,2,2,3,3,4,4,5-d9, Bromo-cyclopentane-d9, Cyclopentyl-d9 Bromide
InChI formel InChI=1S/C5H9Br/c6-5-3-1-2-4-5/h5H,1-4H2/i1D2,2D2,3D2,4D2,5D
SMIL [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(Br)C1([2H])[2H]
IUPAC navn 1-bromo-1,2,2,3,3,4,4,5,5-nonadeuteriocyclopentane
Molekylvægt (g/mol) 158.08
Formel vægt 157.0453