Få mere at vide
Bromocyclopentane-d9, CDN
Artikelnummer. 30323897
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30323897 1 g -
30315292 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323897 Leverandør CDN Leverandørnr. CDND11891G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bromocyclopentane-d9
CAS 35468-44-1
Molekylær formel C5 D9 Br
Mængde 1 g
Synonym 5-Bromo-cyclopentane-1,1,2,2,3,3,4,4,5-d9, Bromo-cyclopentane-d9, Cyclopentyl-d9 Bromide
InChI formel InChI=1 S/C5H9Br/c6-5-3-1-2-4-5/h5H,1-4H2/i1D2,2D2,3D2,4D2,5 D
SMIL [2 H]C1([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(Br)C1([2 H])[2 H]
IUPAC navn 1-bromo-1,2,2,3,3,4,4,5,5-nonadeuteriocyclopentane
Molekylvægt (g/mol) 158.08
Formel vægt 157.0453 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity