Få mere at vide
Bromobenzene-2,3,5,6-d4, CDN
Artikelnummer. 30318412
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318412 1 g
30315407 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318412 Leverandør CDN Leverandørnr. CDND68321G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bromobenzene-2,3,5,6-D4
CAS 64646-03-3
Molekylær formel C6 D4 H Br
Mængde 1 g
Synonym Benzene-1,2,4,5-d4, 3-bromo- (9 CI), Bromobenzene-2,3,5,6-d4, Bromobenzene-2,3,5,6-D4, Bromobenzene-d4, 3-Bromobenzene-1,2,4,5-d4
InChI formel InChI=1 S/C6H5Br/c7-6-4-2-1-3-5-6/h1-5 H/i2D,3 D,4 D,5 D
SMIL [2 H]c1cc([2 H])c([2 H])c(Br)c1[2 H]
IUPAC navn 3-bromo-1,2,4,5-tetradeuteriobenzene
Molekylvægt (g/mol) 161.0326
Formel vægt 159.9826 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity