Få mere at vide
Bromoacetic Acid-d2, TRC
Artikelnummer. 30333113
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30333113 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30333113 Leverandør Toronto Research Chemicals Leverandørnr. TRCB679082100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bromoacetic acid d2
CAS 41830-86-8
Molekylær formel C2 D2 H Br O2
Mængde 100 mg
Synonym Acetic-d2 acid, bromo- (9CI), 2-Bromoacetic-2,2-d2 acid, Bromoacetic Acid-D2, Bromoacetic-d2 acid
InChI formel InChI=1S/C2H3BrO2/c3-1-2(4)5/h1H2,(H,4,5)/i1D2
SMIL [2H]C([2H])(Br)C(=O)O
IUPAC navn 2-bromo-2,2-dideuterio-acetic acid
Molekylvægt (g/mol) 140.96
Formel vægt 139.9442