missing translation for 'onlineSavingsMsg'
Learn More

Bromhexine N-Oxide, Mikromol™

Product Code. 17999804
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
17999804 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17999804 Supplier Mikromol Supplier No. MM0065.09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Bromhexine N-Oxide
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Bromhexine N-Oxide
CAS 2724689-57-8
Analyt- eller komponentnavne Bromhexine N-Oxide
Forsendelsestilstand Room Temperature
Molekylær formel C14 H20 Br2 N2 O
Certificeringer/compliance ISO 17025
Synonym Bromhexine N-Oxide
InChI formel InChI=1S/C14H20Br2N2O/c1-18(19,12-5-3-2-4-6-12)9-10-7-11(15)8-13(16)14(10)17/h7-8,12H,2-6,9,17H2,1H3
SMIL C[N+]([O-])(Cc1cc(Br)cc(Br)c1N)C2CCCCC2
IUPAC navn N-[(2-amino-3,5-dibromophenyl)methyl]-N-methylcyclohexanamine oxide
Molekylvægt (g/mol) 392.13
Mængde 100 mg
Formel vægt 389.9942 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.