Få mere at vide
Brombuterol-d9 Hydrochloride, TRC
Artikelnummer. 30342846
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30342846 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30342846 Leverandør Toronto Research Chemicals Leverandørnr. TRCB67853710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Brombuterol-d9 hydrochloride
CAS 1353867-94-3
Renhedsgrad noter HPLC
Molekylær formel C12 2H9 H9 Br2 N2 O . Cl H
Mængde 10 mg
Synonym 1-(4-Amino-3,5-dibromophenyl)-2-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]ethanol hydrochloride
InChI formel InChI=1S/C12H18Br2N2O.ClH/c1-12(2,3)16-6-10(17)7-4-8(13)11(15)9(14)5-7;/h4-5,10,16-17H,6,15H2,1-3H3;1H/i1D3,2D3,3D3;
SMIL Cl.[2H]C([2H])([2H])C(NCC(O)c1cc(Br)c(N)c(Br)c1)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1-(4-amino-3,5-dibromophenyl)-2-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]ethanol;hydrochloride
Molekylvægt (g/mol) 411.61
Formel vægt 409.0118
Procent renhed >95